The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024848.2 | NP_079124.1 | 12 - 13 | 1471 | 147 |
XM_005244798.2 | XP_005244855.1 | 11 - 12 | 1335 | 147 | |
XM_011542169.1 | XP_011540471.1 | 12 - 13 | 1468 | 147 | |
XM_011542176.1 | XP_011540478.1 | 10 - 11 | 1180 | 147 | |
GenBank mRNA | AK024003.1 | - | 12 - 13 | 1274 | 147 |