The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079670.2 | NP_001073138.1 | 4 - 5 | 495 | 92 |
NM_001287337.1 | NP_001274266.1 | 5 - 6 | 633 | 92 | |
NM_001287338.1 | NP_001274267.1 | 5 - 6 | 628 | 92 | |
NM_001287339.1 | NP_001274268.1 | 3 - 4 | 297 | 92 | |
NM_030925.3 | NP_112187.2 | 2 - 3 | 546 | 92 | |
XM_011535254.2 | XP_011533556.1 | 6 - 7 | 1037 | 92 | |
XM_011535255.2 | XP_011533557.1 | 4 - 5 | 517 | 92 | |
XM_011535256.2 | XP_011533558.1 | 5 - 6 | 782 | 92 | |
XM_017020785.1 | XP_016876274.1 | 5 - 6 | 909 | 92 | |
XM_017020786.1 | XP_016876275.1 | 6 - 7 | 1046 | 92 | |
XM_017020787.1 | XP_016876276.1 | 6 - 7 | 1299 | 92 | |
XM_017020788.1 | XP_016876277.1 | 7 - 8 | 1175 | 92 | |
GenBank mRNA | AK022639.1 | - | 1 - 2 | 91 | 92 |
AY211923.1 | - | 4 - 5 | 408 | 92 | |
AY288977.2 | - | 5 - 6 | 560 | 92 | |
BC010993.2 | - | 2 - 3 | 371 | 92 | |
BX647518.1 | - | 4 - 5 | 432 | 92 | |
DB457246.1 | - | 3 - 4 | 297 | 92 |