The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031889.2 | NP_114095.2 | 5 - 6 | 491 | 89 |
XM_006714056.3 | XP_006714119.1 | 4 - 5 | 2240 | 89 | |
GenBank mRNA | AF125373.1 | - | 5 - 6 | 491 | 89 |
BC117308.1 | - | 4 - 5 | 280 | 89 | |
BC117310.1 | - | 4 - 5 | 280 | 89 |