The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032890.3 | NP_116279.2 | 8 - 9 | 1217 | 81 |
XM_005273335.2 | XP_005273392.1 | 6 - 7 | 1058 | 81 | |
XM_006711592.2 | XP_006711655.1 | 7 - 8 | 2005 | 81 | |
XM_006711594.3 | XP_006711657.1 | 6 - 7 | 713 | 81 | |
XM_011510072.2 | XP_011508374.1 | 7 - 8 | 1057 | 81 | |
XM_011510073.2 | XP_011508375.1 | 7 - 8 | 1535 | 81 | |
XM_011510074.2 | XP_011508376.1 | 7 - 8 | 1076 | 81 | |
XM_011510075.2 | XP_011508377.1 | 7 - 8 | 1563 | 81 | |
XM_011510077.2 | XP_011508379.1 | 6 - 7 | 999 | 81 | |
XM_017002611.1 | XP_016858100.1 | 6 - 7 | 1473 | 81 | |
XM_017002612.1 | XP_016858101.1 | 7 - 8 | 882 | 81 | |
XM_017002613.1 | XP_016858102.1 | 5 - 6 | 842 | 81 | |
GenBank mRNA | AK098669.1 | - | 6 - 7 | 951 | 81 |
AK125728.1 | - | 6 - 7 | 1055 | 81 | |
BC007734.2 | - | 6 - 7 | 1038 | 81 |