The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015380.4 | NP_056195.3 | 7 - 8 | 841 | 64 |
GenBank mRNA | AF151809.1 | - | 7 - 8 | 753 | 64 |
AK001087.1 | - | 7 - 8 | 780 | 64 | |
AK222657.1 | - | 7 - 8 | 715 | 64 | |
AK297770.1 | - | 6 - 7 | 524 | 64 | |
AK312773.1 | - | 7 - 8 | 676 | 64 | |
AY189688.1 | - | 7 - 8 | 756 | 64 | |
BC007830.1 | - | 7 - 8 | 748 | 64 | |
BC011681.2 | - | 7 - 8 | 801 | 64 | |
BC015200.1 | - | 7 - 8 | 691 | 64 | |
CR456483.1 | - | 7 - 8 | 653 | 64 |