The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016052.3 | NP_057136.2 | 3 - 4 | 536 | 133 |
XM_011509597.2 | XP_011507899.1 | 3 - 4 | 553 | 133 | |
GenBank mRNA | AF151873.1 | - | 3 - 4 | 471 | 133 |
AK290066.1 | - | 3 - 4 | 518 | 133 | |
BC020641.1 | - | 3 - 4 | 513 | 133 |