Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MUTYH
Entrez Gene ID
4595
Gene Name
mutY DNA glycosylase
Gene Aliases
MYH
Chromosome Location
Chr.1: 45329242 - 45340925 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048171.1 NP_001041636.1 1 - 2 250 97
NM_001128425.1 NP_001121897.1 1 - 2 250 97
NM_001293190.1 NP_001280119.1 1 - 2 250 97
NM_001293192.1 NP_001280121.1 1 - 2 250 97
NM_012222.2 NP_036354.1 1 - 2 250 97
XM_011541503.2 XP_011539805.1 1 - 2 95 97
XM_011541505.2 XP_011539807.1 1 - 2 101 97
XM_017001336.1 XP_016856825.1 1 - 2 145 97
XM_017001338.1 XP_016856827.1 1 - 2 705 97
XM_017001339.1 XP_016856828.1 1 - 2 220 97
GenBank mRNA AB032920.1 - 1 - 2 168 97
AB032921.1 - 1 - 2 168 97
AB032922.1 - 1 - 2 168 97
AB032923.1 - 1 - 2 168 97
AK293830.1 - 1 - 2 227 97
BC003178.1 - 1 - 2 188 97
BM926983.1 - 1 - 2 250 97

View Full Product Details