Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN11
Entrez Gene ID
91544
Gene Name
UBX domain protein 11
Gene Aliases
COA-1, PP2243, SOC, SOCI, UBXD5
Chromosome Location
Chr.1: 26282282 - 26318265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY007163.1 - 5 - 6 291 100

View Full Product Details