Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AXDND1
Entrez Gene ID
126859
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
C1orf125
Chromosome Location
Chr.1: 179365705 - 179554735 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144696.5 NP_653297.3 8 - 9 1127 90
NR_073544.1 8 - 9 1001 90
XM_011509165.1 XP_011507467.1 8 - 9 1106 90
XM_011509166.2 XP_011507468.1 8 - 9 1153 90
XM_011509167.2 XP_011507469.1 8 - 9 1268 90
XM_011509168.1 XP_011507470.1 8 - 9 1110 90
XM_011509169.1 XP_011507471.1 7 - 8 899 90
XM_011509170.1 XP_011507472.1 8 - 9 1106 90
XM_011509171.1 XP_011507473.1 7 - 8 924 90
XM_011509174.1 XP_011507476.1 8 - 9 1106 90
XM_011509175.1 XP_011507477.1 8 - 9 1106 90
XM_011509176.1 XP_011507478.1 8 - 9 1106 90
XM_011509178.2 XP_011507480.1 8 - 9 1106 90
XM_011509179.2 XP_011507481.1 3 - 4 214 90
XM_011509180.1 XP_011507482.1 8 - 9 1106 90
GenBank mRNA AK057502.1 - 6 - 7 494 90
AK302972.1 - 8 - 9 1106 90
AL833914.1 - 6 - 7 554 90
BC146823.1 - 8 - 9 1002 90
BX647935.1 - 8 - 9 1128 90

View Full Product Details