Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72352983-72353480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
UniGene
Hs.604479
Gene Chromosome Location
Chr.15: 72343435-72376473 on Build GRCh38
Transcripts
NM_000520.5 NM_001318825.1 NR_134869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3401904 c.509G>A p.R170Q Chr.15: 72353129-72353129 on Build GRCh38
471030 c.559C>A p.L187M Chr.15: 72353079-72353079 on Build GRCh38
6274551 c.496C>A p.R166S Chr.15: 72353142-72353142 on Build GRCh38
4500150 c.558C>T p.I186I Chr.15: 72353080-72353080 on Build GRCh38
7277247 c.497G>A p.R166H Chr.15: 72353141-72353141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005565912 Chr.15: 72353327-72353327 on Build GRCh38
rs531453216 Chr.15: 72353032-72353032 on Build GRCh38
rs771586204 Chr.15: 72353216-72353216 on Build GRCh38
rs372532840 Chr.15: 72353226-72353226 on Build GRCh38
rs905274037 Chr.15: 72353361-72353361 on Build GRCh38
rs761444674 Chr.15: 72353282-72353282 on Build GRCh38
rs185797496 Chr.15: 72353090-72353090 on Build GRCh38
rs28941770 Chr.15: 72353105-72353105 on Build GRCh38
rs556888199 Chr.15: 72353256-72353256 on Build GRCh38
rs776464763 Chr.15: 72353229-72353229 on Build GRCh38
rs28941771 Chr.15: 72353100-72353100 on Build GRCh38
rs1036054167 Chr.15: 72353010-72353010 on Build GRCh38
rs781449664 Chr.15: 72353043-72353045 on Build GRCh38
rs781493030 Chr.15: 72353103-72353103 on Build GRCh38
rs904857940 Chr.15: 72353235-72353235 on Build GRCh38
rs896096863 Chr.15: 72353308-72353308 on Build GRCh38
rs752813753 Chr.15: 72353166-72353166 on Build GRCh38
rs764343937 Chr.15: 72353179-72353179 on Build GRCh38
rs987519840 Chr.15: 72353437-72353437 on Build GRCh38
rs777360331 Chr.15: 72353217-72353217 on Build GRCh38
rs760533263 Chr.15: 72353038-72353038 on Build GRCh38
rs570948892 Chr.15: 72353457-72353457 on Build GRCh38
rs747372270 Chr.15: 72353102-72353102 on Build GRCh38
rs368112859 Chr.15: 72353024-72353024 on Build GRCh38
rs946429436 Chr.15: 72353181-72353181 on Build GRCh38
rs771696420 Chr.15: 72353119-72353119 on Build GRCh38
rs796165682 Chr.15: 72353355-72353355 on Build GRCh38
rs121907957 Chr.15: 72353129-72353129 on Build GRCh38
rs960250447 Chr.15: 72353421-72353421 on Build GRCh38
rs746698871 Chr.15: 72353221-72353221 on Build GRCh38
rs764627152 Chr.15: 72353311-72353311 on Build GRCh38
rs753831831 Chr.15: 72353072-72353072 on Build GRCh38
rs759487463 Chr.15: 72353142-72353142 on Build GRCh38
rs535348939 Chr.15: 72353364-72353364 on Build GRCh38
rs190992885 Chr.15: 72353373-72353373 on Build GRCh38
rs1057519460 Chr.15: 72353114-72353114 on Build GRCh38
rs144652159 Chr.15: 72353080-72353080 on Build GRCh38
rs758618617 Chr.15: 72353174-72353174 on Build GRCh38
rs930231608 Chr.15: 72353169-72353169 on Build GRCh38
rs148511084 Chr.15: 72353093-72353093 on Build GRCh38
rs766165730 Chr.15: 72353062-72353062 on Build GRCh38
rs370829895 Chr.15: 72353200-72353200 on Build GRCh38
rs568565456 Chr.15: 72353372-72353372 on Build GRCh38
rs786204754 Chr.15: 72353067-72353067 on Build GRCh38
rs761481637 Chr.15: 72353020-72353020 on Build GRCh38
rs988910765 Chr.15: 72353012-72353012 on Build GRCh38
rs121907953 Chr.15: 72353106-72353106 on Build GRCh38
rs759310073 Chr.15: 72353230-72353230 on Build GRCh38
rs200743199 Chr.15: 72353121-72353121 on Build GRCh38
rs760406474 Chr.15: 72353128-72353128 on Build GRCh38
rs1057499783 Chr.15: 72353298-72353298 on Build GRCh38
rs765136263 Chr.15: 72353156-72353156 on Build GRCh38
rs373914061 Chr.15: 72353207-72353207 on Build GRCh38
rs529900628 Chr.15: 72353199-72353199 on Build GRCh38
rs371576105 Chr.15: 72353040-72353040 on Build GRCh38
rs547177633 Chr.15: 72353180-72353180 on Build GRCh38
rs754966985 Chr.15: 72353074-72353074 on Build GRCh38
rs12440768 Chr.15: 72353011-72353011 on Build GRCh38
rs147090337 Chr.15: 72353416-72353416 on Build GRCh38
rs1015055430 Chr.15: 72353336-72353336 on Build GRCh38
rs1045141526 Chr.15: 72353345-72353345 on Build GRCh38
rs121907972 Chr.15: 72353130-72353130 on Build GRCh38
rs1003199951 Chr.15: 72353261-72353261 on Build GRCh38
rs780593404 Chr.15: 72353204-72353204 on Build GRCh38
rs1057516850 Chr.15: 72353173-72353173 on Build GRCh38
rs200177553 Chr.15: 72353195-72353195 on Build GRCh38
rs912878006 Chr.15: 72353021-72353021 on Build GRCh38
rs1035514333 Chr.15: 72353410-72353410 on Build GRCh38
rs121907969 Chr.15: 72353098-72353098 on Build GRCh38
rs200513831 Chr.15: 72353099-72353099 on Build GRCh38
rs911398032 Chr.15: 72353087-72353087 on Build GRCh38
rs75174831 Chr.15: 72353104-72353104 on Build GRCh38
rs747703708 Chr.15: 72353107-72353107 on Build GRCh38
rs953340351 Chr.15: 72353400-72353400 on Build GRCh38
rs398123442 Chr.15: 72353141-72353141 on Build GRCh38

View Full Product Details