The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001223.4 | NP_001214.1 | 7 - 8 | 1099 | 76 |
NM_001257118.2 | NP_001244047.1 | 8 - 9 | 1162 | 76 | |
NM_001257119.2 | NP_001244048.1 | 7 - 8 | 1099 | 76 | |
NM_033292.3 | NP_150634.1 | 8 - 9 | 1162 | 76 | |
NM_033293.3 | NP_150635.1 | 7 - 8 | 883 | 76 | |
NM_033294.3 | NP_150636.1 | 6 - 7 | 739 | 76 | |
NM_033295.3 | NP_150637.1 | 3 - 4 | 214 | 76 | |
XM_017018393.1 | XP_016873882.1 | 8 - 9 | 1510 | 76 | |
XM_017018394.1 | XP_016873883.1 | 8 - 9 | 1510 | 76 | |
XM_017018395.1 | XP_016873884.1 | 7 - 8 | 1447 | 76 | |
XM_017018396.1 | XP_016873885.1 | 8 - 9 | 1419 | 76 | |
GenBank mRNA | AA761907.1 | - | 2 - 3 | 76 | |
AK223503.1 | - | 8 - 9 | 1154 | 76 | |
AK290114.1 | - | 8 - 9 | 1153 | 76 | |
AK290122.1 | - | 7 - 8 | 1090 | 76 | |
AK313516.1 | - | 8 - 9 | 1148 | 76 | |
AY660536.1 | - | 8 - 9 | 1056 | 76 | |
BC041689.2 | - | 6 - 7 | 1048 | 76 | |
BC062327.1 | - | 7 - 8 | 849 | 76 | |
M87507.1 | - | 8 - 9 | 1135 | 76 | |
U13697.1 | - | 6 - 7 | 1055 | 76 | |
U13698.1 | - | 6 - 7 | 839 | 76 | |
U13699.1 | - | 5 - 6 | 695 | 76 | |
U13700.1 | - | 2 - 3 | 170 | 76 | |
X65019.1 | - | 7 - 8 | 1125 | 76 |