The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002111.8 | NP_002102.4 | 17 - 18 | 2544 | 77 |
GenBank mRNA | AB016794.1 | - | 17 - 18 | 2544 | 77 |
AB209506.1 | - | 4 - 5 | 2492 | 77 | |
L12392.1 | - | 17 - 18 | 2714 | 77 |