Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NEUROG3
Entrez Gene ID
50674
Gene Name
neurogenin 3
Gene Aliases
Atoh5, Math4B, NGN-3, bHLHa7, ngn3
Chromosome Location
Chr.10: 69571440 - 69573454 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020999.3 NP_066279.2 1 - 2 243 83
GenBank mRNA AK313952.1 - 1 - 2 246 83
BC117488.1 - 1 - 2 82 83
BC126468.1 - 1 - 2 82 83

View Full Product Details