Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NYX
Entrez Gene ID
60506
Gene Name
nyctalopin
Gene Aliases
CLRP, CSNB1, CSNB1A, CSNB4, NBM1
Chromosome Location
Chr.X: 41447460 - 41476414 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022567.2 NP_072089.1 1 - 2 474 91
XM_017029709.1 XP_016885198.1 2 - 3 142 91
GenBank mRNA AF254868.1 - 2 - 3 142 91
AJ278865.1 - 1 - 2 500 91
BC112242.1 - 1 - 2 88 91

View Full Product Details