The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001309192.1 | NP_001296121.1 | 5 - 6 | 1448 | 72 |
NM_001309193.1 | NP_001296122.1 | 5 - 6 | 1448 | 72 | |
NM_004442.7 | NP_004433.2 | 5 - 6 | 1448 | 72 | |
NM_017449.4 | NP_059145.2 | 5 - 6 | 1448 | 72 | |
XM_006710441.3 | XP_006710504.1 | 5 - 6 | 1541 | 72 | |
XM_006710442.3 | XP_006710505.1 | 5 - 6 | 1516 | 72 | |
GenBank mRNA | AB210018.1 | - | 5 - 6 | 1357 | 72 |
AF025304.1 | - | 5 - 6 | 1328 | 72 | |
AK298898.1 | - | 1 - 2 | 125 | 72 | |
AK299701.1 | - | 6 - 7 | 1387 | 72 | |
BC007903.2 | - | 4 - 5 | 1226 | 72 | |
BC018763.1 | - | 3 - 4 | 601 | 72 | |
BC067861.1 | - | 5 - 6 | 1447 | 72 | |
D31661.1 | - | 5 - 6 | 1309 | 72 | |
L36643.1 | - | 5 - 6 | 1261 | 72 | |
L41939.1 | - | 5 - 6 | 1407 | 72 |