The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317950.1 | NP_001304879.1 | 12 - 13 | 2955 | 95 |
NM_001317952.1 | NP_001304881.1 | 11 - 12 | 2513 | 95 | |
NM_030767.5 | NP_110394.3 | 12 - 13 | 2900 | 95 | |
XM_005252244.2 | XP_005252301.1 | 14 - 15 | 3072 | 95 | |
XM_005252245.1 | XP_005252302.1 | 13 - 14 | 2930 | 95 | |
XM_005252247.4 | XP_005252304.1 | 12 - 13 | 2951 | 95 | |
XM_005252248.2 | XP_005252305.1 | 12 - 13 | 3068 | 95 | |
XM_006717294.1 | XP_006717357.1 | 12 - 13 | 2884 | 95 | |
XM_011519063.2 | XP_011517365.1 | 11 - 12 | 2563 | 95 | |
XM_011519064.2 | XP_011517366.1 | 12 - 13 | 3068 | 95 | |
XM_011519065.2 | XP_011517367.2 | 12 - 13 | 2543 | 95 | |
XM_017015172.1 | XP_016870661.1 | 13 - 14 | 2670 | 95 | |
GenBank mRNA | AF286341.1 | - | 2 - 3 | 455 | 95 |
AK024431.1 | - | 10 - 11 | 1848 | 95 | |
AK074040.1 | - | 11 - 12 | 1943 | 95 | |
AK131082.2 | - | 12 - 13 | 2955 | 95 | |
AK160382.1 | - | 12 - 13 | 2956 | 95 | |
AL833220.1 | - | 9 - 10 | 1377 | 95 | |
AY703039.1 | - | 13 - 14 | 2555 | 95 | |
AY703040.1 | - | 11 - 12 | 2513 | 95 | |
AY703043.1 | - | 11 - 12 | 2873 | 95 | |
AY703044.1 | - | 10 - 11 | 2567 | 95 | |
AY703045.1 | - | 3 - 4 | 650 | 95 | |
BC055285.1 | - | 12 - 13 | 2900 | 95 |