Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKNA
Entrez Gene ID
80709
Gene Name
AT-hook transcription factor
Gene Aliases
-
Chromosome Location
Chr.9: 114334149 - 114398498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317950.1 NP_001304879.1 19 - 20 3976 54
NM_001317952.1 NP_001304881.1 18 - 19 3534 54
NM_030767.5 NP_110394.3 19 - 20 3921 54
XM_005252244.2 XP_005252301.1 21 - 22 4093 54
XM_005252245.1 XP_005252302.1 20 - 21 3951 54
XM_005252247.4 XP_005252304.1 19 - 20 3972 54
XM_006717294.1 XP_006717357.1 19 - 20 3905 54
XM_011519063.2 XP_011517365.1 18 - 19 3584 54
XM_011519065.2 XP_011517367.2 19 - 20 3564 54
XM_017015172.1 XP_016870661.1 20 - 21 3691 54
GenBank mRNA AF286341.1 - 9 - 10 1475 54
AK024431.1 - 17 - 18 2869 54
AK074040.1 - 18 - 19 2964 54
AK131082.2 - 19 - 20 3976 54
AK160382.1 - 19 - 20 3978 54
AL713755.1 - 2 - 3 2410 54
AL833220.1 - 16 - 17 2398 54
AY703039.1 - 20 - 21 3576 54
AY703040.1 - 18 - 19 3534 54
AY703043.1 - 18 - 19 3894 54
AY703044.1 - 17 - 18 3588 54
AY703045.1 - 10 - 11 1667 54
AY703046.1 - 6 - 7 793 54
BC042202.1 - 5 - 6 651 54
BC055285.1 - 19 - 20 3921 54

View Full Product Details