Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF14
Entrez Gene ID
9604
Gene Name
ring finger protein 14
Gene Aliases
ARA54, HFB30, HRIHFB2038, TRIAD2
Chromosome Location
Chr.5: 141949337 - 141993709 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201365.1 NP_001188294.1 2 - 3 241 89
NM_004290.4 NP_004281.1 2 - 3 455 89
NM_183398.2 NP_899645.1 2 - 3 455 89
NM_183401.2 NP_899648.1 2 - 3 261 89
XM_005268536.2 XP_005268593.1 2 - 3 558 89
XM_005268537.2 XP_005268594.1 2 - 3 602 89
XM_005268539.2 XP_005268596.1 2 - 3 223 89
XM_005268540.3 XP_005268597.1 2 - 3 379 89
XM_005268542.2 XP_005268599.1 2 - 3 298 89
XM_017010070.1 XP_016865559.1 2 - 3 206 89
XM_017010071.1 XP_016865560.1 2 - 3 585 89
XM_017010072.1 XP_016865561.1 2 - 3 545 89
XM_017010073.1 XP_016865562.1 2 - 3 336 89
XM_017010074.1 XP_016865563.1 2 - 3 602 89
XM_017010075.1 XP_016865564.1 2 - 3 261 89
XM_017010076.1 XP_016865565.1 2 - 3 247 89
GenBank mRNA AB022663.1 - 2 - 3 229 89
AK023884.1 - 2 - 3 197 89
AK057868.1 - 2 - 3 231 89
AK129515.1 - 2 - 3 384 89
AK294262.1 - 2 - 3 200 89
AK295965.1 - 2 - 3 250 89
AW162926.1 - 2 - 3 284 89
BC126185.1 - 2 - 3 200 89
BC144061.1 - 2 - 3 200 89
BX647955.1 - 2 - 3 228 89
CD106708.1 - 2 - 3 267 89
DB456366.2 - 2 - 3 298 89
DC358984.1 - 2 - 3 261 89
DC396586.1 - 2 - 3 241 89
DC402536.1 - 2 - 3 458 89

View Full Product Details