Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTFMT
Entrez Gene ID
123263
Gene Name
mitochondrial methionyl-tRNA formyltransferase
Gene Aliases
COXPD15, FMT1
Chromosome Location
Chr.15: 65001512 - 65029639 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139242.3 NP_640335.2 3 - 4 563 79
XM_005254158.4 XP_005254215.2 3 - 4 705 79
GenBank mRNA AK055688.1 - 3 - 4 509 79
BC016630.1 - 3 - 4 541 79
BC033687.1 - 3 - 4 551 79

View Full Product Details