The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144611.3 | NP_653212.1 | 1 - 2 | 842 | 59 |
XM_017024183.1 | XP_016879672.1 | 1 - 2 | 536 | 59 | |
GenBank mRNA | AK172844.1 | - | 1 - 2 | 308 | 59 |
AK313088.1 | - | 1 - 2 | 313 | 59 | |
BC020263.1 | - | 1 - 2 | 294 | 59 | |
BC051697.1 | - | 1 - 2 | 754 | 59 |