Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGI2
Entrez Gene ID
55203
Gene Name
leucine rich repeat LGI family member 2
Gene Aliases
LGIL2
Chromosome Location
Chr.4: 24991960 - 25030792 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018176.3 NP_060646.2 1 - 2 299 62
XM_011513850.2 XP_011512152.1 1 - 2 201 62
XM_017008356.1 XP_016863845.1 1 - 2 201 62
GenBank mRNA AB067503.1 - 1 - 2 192 62
AF467955.1 - 1 - 2 200 62
AJ487516.1 - 1 - 2 200 62
AJ487958.1 - 1 - 2 222 62
BC101759.1 - 1 - 2 299 62
BC101761.1 - 1 - 2 299 62

View Full Product Details