The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134405.1 | NP_001127877.1 | 6 - 7 | 1036 | 100 |
NM_001134406.1 | NP_001127878.1 | 6 - 7 | 1036 | 100 | |
NM_138290.2 | NP_612147.1 | 7 - 8 | 1087 | 100 | |
XM_005250156.2 | XP_005250213.1 | 6 - 7 | 1036 | 100 | |
XM_005250158.2 | XP_005250215.1 | 4 - 5 | 511 | 100 | |
XM_011515826.1 | XP_011514128.1 | 7 - 8 | 1087 | 100 | |
XM_011515827.1 | XP_011514129.1 | 7 - 8 | 1087 | 100 | |
XM_011515828.2 | XP_011514130.1 | 7 - 8 | 570 | 100 | |
XM_011515829.2 | XP_011514131.1 | 6 - 7 | 704 | 100 | |
XM_017011761.1 | XP_016867250.1 | 7 - 8 | 774 | 100 | |
XM_017011762.1 | XP_016867251.1 | 8 - 9 | 699 | 100 | |
XM_017011763.1 | XP_016867252.1 | 5 - 6 | 513 | 100 | |
XM_017011764.1 | XP_016867253.1 | 4 - 5 | 511 | 100 | |
XM_017011765.1 | XP_016867254.1 | 7 - 8 | 686 | 100 | |
XM_017011766.1 | XP_016867255.1 | 7 - 8 | 965 | 100 | |
GenBank mRNA | AK055233.1 | - | 7 - 8 | 1085 | 100 |
AK293607.1 | - | 6 - 7 | 852 | 100 | |
AK294039.1 | - | 6 - 7 | 834 | 100 | |
AY033596.1 | - | 5 - 6 | 522 | 100 | |
BC022520.1 | - | 6 - 7 | 987 | 100 |