Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WWC3
Entrez Gene ID
55841
Gene Name
WWC family member 3
Gene Aliases
BM042
Chromosome Location
Chr.X: 10015755 - 10144478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015691.3 NP_056506.2 16 - 17 2477 71
GenBank mRNA AB033106.1 - 14 - 15 2100 71
AK024683.1 - 7 - 8 1287 71
AK091936.1 - 7 - 8 1154 71
AK292961.1 - 15 - 16 2359 71
AL353937.1 - 13 - 14 1968 71
BC032447.1 - 13 - 14 2012 71
KC987947.1 - 16 - 17 2654 71

View Full Product Details