Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPH7
Entrez Gene ID
92715
Gene Name
diphthamide biosynthesis 7
Gene Aliases
C9orf112, RRT2, WDR85
Chromosome Location
Chr.9: 137554899 - 137578935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138778.3 NP_620133.1 1 - 2 309 91
XM_005266123.2 XP_005266180.1 1 - 2 299 91
XM_005266128.2 XP_005266185.1 1 - 2 299 91
XM_005266129.2 XP_005266186.1 1 - 2 299 91
XM_011519188.1 XP_011517490.1 1 - 2 298 91
XM_011519190.2 XP_011517492.1 1 - 2 298 91
XM_011519191.2 XP_011517493.1 1 - 2 298 91
XM_011519192.2 XP_011517494.1 1 - 2 298 91
XM_011519193.2 XP_011517495.1 1 - 2 298 91
XM_011519194.1 XP_011517496.1 1 - 2 298 91
XM_017015289.1 XP_016870778.1 1 - 2 298 91
XM_017015290.1 XP_016870779.1 1 - 2 298 91
XM_017015292.1 XP_016870781.1 1 - 2 298 91
XM_017015294.1 XP_016870783.1 1 - 2 299 91
XM_017015295.1 XP_016870784.1 1 - 2 298 91
XM_017015297.1 XP_016870786.1 1 - 2 299 91
XM_017015298.1 XP_016870787.1 1 - 2 299 91
XM_017015300.1 XP_016870789.1 1 - 2 299 91
XM_017015303.1 XP_016870792.1 1 - 2 298 91
XM_017015305.1 XP_016870794.1 1 - 2 299 91
XM_017015306.1 XP_016870795.1 1 - 2 299 91
XM_017015307.1 XP_016870796.1 1 - 2 299 91
XM_017015309.1 XP_016870798.1 1 - 2 299 91
GenBank mRNA AK075115.1 - 1 - 2 310 91
BC003123.1 - 1 - 2 248 91
BC017335.2 - 1 - 2 268 91

View Full Product Details