The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147128.3 | NP_667339.1 | 3 - 4 | 1719 | 82 |
GenBank mRNA | AF513707.1 | - | 3 - 4 | 1719 | 82 |
AF527533.1 | - | 3 - 4 | 1046 | 82 | |
AK025240.1 | - | 2 - 3 | 161 | 82 | |
AY054122.1 | - | 4 - 5 | 483 | 82 |