Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPRD2
Entrez Gene ID
23248
Gene Name
regulation of nuclear pre-mRNA domain containing 2
Gene Aliases
HSPC099, KIAA0460
Chromosome Location
Chr.1: 150364111 - 150476566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297673.1 NP_001284602.1 4 - 5 1093 86
NM_001297674.1 NP_001284603.1 4 - 5 1093 86
NM_015203.4 NP_056018.2 5 - 6 1171 86
XM_005245033.2 XP_005245090.1 5 - 6 1134 86
XM_006711240.2 XP_006711303.1 5 - 6 1134 86
XM_011509344.2 XP_011507646.1 5 - 6 1134 86
XM_017000763.1 XP_016856252.1 4 - 5 1056 86
XM_017000764.1 XP_016856253.1 5 - 6 509 86
XM_017000765.1 XP_016856254.1 5 - 6 1134 86
XM_017000766.1 XP_016856255.1 4 - 5 768 86
XM_017000767.1 XP_016856256.1 4 - 5 1056 86
GenBank mRNA AB007929.2 - 5 - 6 540 86
AK056707.1 - 5 - 6 578 86
AK125864.1 - 5 - 6 741 86
AK291703.1 - 5 - 6 589 86
AK304380.1 - 4 - 5 511 86
AK308148.1 - 4 - 5 502 86
BC045623.1 - 4 - 5 494 86
BX641025.1 - 6 - 7 1066 86

View Full Product Details