Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPRD2
Entrez Gene ID
23248
Gene Name
regulation of nuclear pre-mRNA domain containing 2
Gene Aliases
HSPC099, KIAA0460
Chromosome Location
Chr.1: 150364111 - 150476566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297673.1 NP_001284602.1 9 - 10 2138 77
NM_001297674.1 NP_001284603.1 9 - 10 2138 77
NM_015203.4 NP_056018.2 10 - 11 2216 77
XM_005245033.2 XP_005245090.1 10 - 11 2179 77
XM_006711240.2 XP_006711303.1 10 - 11 2179 77
XM_017000764.1 XP_016856253.1 10 - 11 1554 77
XM_017000766.1 XP_016856255.1 9 - 10 1813 77
GenBank mRNA AB007929.2 - 10 - 11 1585 77
AK056707.1 - 10 - 11 1623 77
AK125864.1 - 10 - 11 1786 77
AK291703.1 - 10 - 11 1634 77
AK304380.1 - 9 - 10 1556 77
AK308148.1 - 9 - 10 1547 77
BC045623.1 - 9 - 10 1539 77
BX641025.1 - 11 - 12 2111 77

View Full Product Details