Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKNA
Entrez Gene ID
80709
Gene Name
AT-hook transcription factor
Gene Aliases
-
Chromosome Location
Chr.9: 114334149 - 114398498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317950.1 NP_001304879.1 4 - 5 1636 131
NM_001317952.1 NP_001304881.1 3 - 4 1194 131
NM_030767.5 NP_110394.3 4 - 5 1581 131
XM_005252244.2 XP_005252301.1 6 - 7 1753 131
XM_005252245.1 XP_005252302.1 5 - 6 1611 131
XM_005252247.4 XP_005252304.1 4 - 5 1632 131
XM_005252248.2 XP_005252305.1 4 - 5 1749 131
XM_006717294.1 XP_006717357.1 4 - 5 1565 131
XM_006717295.2 XP_006717358.1 4 - 5 1749 131
XM_011519063.2 XP_011517365.1 3 - 4 1244 131
XM_011519064.2 XP_011517366.1 4 - 5 1749 131
XM_011519065.2 XP_011517367.2 4 - 5 1224 131
XM_011519066.2 XP_011517368.1 4 - 5 1749 131
XM_017015172.1 XP_016870661.1 5 - 6 1351 131
GenBank mRNA AB075848.1 - 4 - 5 1754 131
AK024431.1 - 2 - 3 529 131
AK074040.1 - 2 - 3 577 131
AK131082.2 - 4 - 5 1636 131
AK160382.1 - 4 - 5 1636 131
AL833220.1 - 1 - 2 58 131
AY703039.1 - 5 - 6 1236 131
AY703040.1 - 3 - 4 1194 131
AY703041.1 - 5 - 6 950 131
AY703042.1 - 3 - 4 763 131
AY703043.1 - 3 - 4 1554 131
AY703044.1 - 2 - 3 1248 131
BC055285.1 - 4 - 5 1581 131

View Full Product Details