The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175066.3 | NP_778236.2 | 10 - 11 | 1596 | 92 |
GenBank mRNA | AB593157.1 | - | 10 - 11 | 1616 | 92 |
AK097078.1 | - | 10 - 11 | 1596 | 92 | |
BC012461.2 | - | 1 - 2 | 119 | 92 | |
BC040185.2 | - | 10 - 11 | 1569 | 92 | |
CR936870.1 | - | 2 - 3 | 615 | 92 |