The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153603.3 | NP_705831.1 | 14 - 15 | 2085 | 81 |
XM_017023870.1 | XP_016879359.1 | 14 - 15 | 2128 | 81 | |
GenBank mRNA | AF070568.1 | - | 6 - 7 | 657 | 81 |
AK093989.1 | - | 11 - 12 | 1441 | 81 | |
AY358632.1 | - | 9 - 10 | 1122 | 81 | |
BC000549.2 | - | 14 - 15 | 2040 | 81 | |
BC037563.1 | - | 14 - 15 | 2074 | 81 | |
KU178837.1 | - | 14 - 15 | 1888 | 81 |