Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COG7
Entrez Gene ID
91949
Gene Name
component of oligomeric golgi complex 7
Gene Aliases
CDG2E
Chromosome Location
Chr.16: 23388493 - 23453215 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153603.3 NP_705831.1 16 - 17 2338 64
XM_017023870.1 XP_016879359.1 16 - 17 2381 64
GenBank mRNA AF070568.1 - 8 - 9 910 64
AK093989.1 - 13 - 14 1694 64
AK130714.1 - 3 - 4 2123 64
AY358632.1 - 11 - 12 1375 64
BC000549.2 - 16 - 17 2293 64
BC037563.1 - 16 - 17 2327 64
KU178837.1 - 16 - 17 2141 64

View Full Product Details