The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001624.3 | NP_001615.2 | 1 - 2 | 1188 | 104 |
XM_005266839.3 | XP_005266896.1 | 3 - 4 | 2026 | 104 | |
XM_017010333.1 | XP_016865822.1 | 1 - 2 | 1413 | 104 | |
GenBank mRNA | AK056705.1 | - | 1 - 2 | 1188 | 104 |
AK125137.1 | - | 3 - 4 | 2026 | 104 | |
U83115.1 | - | 1 - 2 | 443 | 104 |