Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ISY1
Entrez Gene ID
57461
Gene Name
ISY1 splicing factor homolog
Gene Aliases
FSAP33
Chromosome Location
Chr.3: 129127416 - 129161230 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199469.1 NP_001186398.1 5 - 6 439 121
NM_020701.3 NP_065752.1 5 - 6 439 121
GenBank mRNA BC004442.1 - 5 - 6 254 121
BC007409.2 - 5 - 6 257 121
BC019849.1 - 5 - 6 395 121
BG282120.1 - 5 - 6 440 121

Gene Symbol
ISY1-RAB43
Entrez Gene ID
100534599
Gene Name
ISY1-RAB43 readthrough
Gene Aliases
-
Chromosome Location
Chr.3: 129087569 - 129161230 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204890.1 NP_001191819.1 5 - 6 439 121
GenBank mRNA AB032986.1 - 5 - 6 245 121

View Full Product Details