Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FAM72A
- Entrez Gene ID
-
729533
- Gene Name
- family with sequence similarity 72 member A
- Gene Aliases
-
LMPIP, Ugene
- Chromosome Location
-
Chr.1: 206186179 - 206204918 on Build GRCh38
- Gene Symbol
- FAM72B
- Entrez Gene ID
-
653820
- Gene Name
- family with sequence similarity 72 member B
- Gene Aliases
-
p17
- Chromosome Location
-
Chr.1: 121167646 - 121186896 on Build GRCh38
- Gene Symbol
- FAM72C
- Entrez Gene ID
-
554282
- Gene Name
- family with sequence similarity 72 member C
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 143955364 - 143974219 on Build GRCh38
- Gene Symbol
- FAM72D
- Entrez Gene ID
-
728833
- Gene Name
- family with sequence similarity 72 member D
- Gene Aliases
-
GCUD2
- Chromosome Location
-
Chr.1: 145093805 - 145112696 on Build GRCh38
View Full Product Details