Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BBOF1
Entrez Gene ID
80127
Gene Name
basal body orientation factor 1
Gene Aliases
C14orf45, CCDC176
Chromosome Location
Chr.14: 74019350 - 74078351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005268098.4 XP_005268155.1 2 - 3 180 123
XM_011537177.2 XP_011535479.1 2 - 3 180 123
XM_017021660.1 XP_016877149.1 2 - 3 192 123
XM_017021661.1 XP_016877150.1 2 - 3 244 123
GenBank mRNA DB484577.1 - 2 - 3 244 123

View Full Product Details