Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72345015-72345596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
UniGene
Hs.604479
Gene Chromosome Location
Chr.15: 72343435-72376473 on Build GRCh38
Transcripts
NM_000520.5 NM_001318825.1 NR_134869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7162978 c.1435G>C p.A479P Chr.15: 72345537-72345537 on Build GRCh38
6142825 c.1502C>T p.S501L Chr.15: 72345470-72345470 on Build GRCh38
4056689 c.1487C>T p.A496V Chr.15: 72345485-72345485 on Build GRCh38
5956208 c.1524G>A p.L508L Chr.15: 72345448-72345448 on Build GRCh38
254720 c.1511G>T p.R504L Chr.15: 72345461-72345461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942097828 Chr.15: 72345337-72345337 on Build GRCh38
rs118002327 Chr.15: 72345338-72345338 on Build GRCh38
rs747528877 Chr.15: 72345560-72345560 on Build GRCh38
rs996081388 Chr.15: 72345256-72345256 on Build GRCh38
rs773338562 Chr.15: 72345511-72345511 on Build GRCh38
rs942212689 Chr.15: 72345564-72345564 on Build GRCh38
rs28942071 Chr.15: 72345462-72345462 on Build GRCh38
rs1020196816 Chr.15: 72345543-72345543 on Build GRCh38
rs1002507303 Chr.15: 72345289-72345289 on Build GRCh38
rs567260742 Chr.15: 72345068-72345068 on Build GRCh38
rs373781108 Chr.15: 72345485-72345485 on Build GRCh38
rs916219265 Chr.15: 72345300-72345300 on Build GRCh38
rs560132087 Chr.15: 72345577-72345577 on Build GRCh38
rs767554113 Chr.15: 72345498-72345498 on Build GRCh38
rs572004811 Chr.15: 72345326-72345326 on Build GRCh38
rs75015614 Chr.15: 72345282-72345282 on Build GRCh38
rs62022857 Chr.15: 72345284-72345284 on Build GRCh38
rs954213080 Chr.15: 72345333-72345333 on Build GRCh38
rs948974391 Chr.15: 72345179-72345179 on Build GRCh38
rs797044433 Chr.15: 72345462-72345462 on Build GRCh38
rs1057519468 Chr.15: 72345518-72345518 on Build GRCh38
rs1003000805 Chr.15: 72345208-72345208 on Build GRCh38
rs1031427462 Chr.15: 72345315-72345315 on Build GRCh38
rs778293061 Chr.15: 72345455-72345455 on Build GRCh38
rs954565361 Chr.15: 72345261-72345261 on Build GRCh38
rs775654868 Chr.15: 72345187-72345187 on Build GRCh38
rs760775455 Chr.15: 72345522-72345522 on Build GRCh38
rs936312158 Chr.15: 72345069-72345069 on Build GRCh38
rs890404594 Chr.15: 72345376-72345376 on Build GRCh38
rs113665670 Chr.15: 72345375-72345378 on Build GRCh38
rs10468051 Chr.15: 72345258-72345258 on Build GRCh38
rs556107005 Chr.15: 72345200-72345200 on Build GRCh38
rs900459035 Chr.15: 72345350-72345350 on Build GRCh38
rs886559976 Chr.15: 72345223-72345223 on Build GRCh38
rs756325812 Chr.15: 72345412-72345412 on Build GRCh38
rs761736583 Chr.15: 72345497-72345497 on Build GRCh38
rs1011316903 Chr.15: 72345320-72345320 on Build GRCh38
rs948670536 Chr.15: 72345236-72345236 on Build GRCh38
rs121907956 Chr.15: 72345476-72345476 on Build GRCh38
rs121907955 Chr.15: 72345461-72345461 on Build GRCh38
rs149092488 Chr.15: 72345575-72345575 on Build GRCh38
rs758829206 Chr.15: 72345548-72345548 on Build GRCh38
rs772240416 Chr.15: 72345349-72345349 on Build GRCh38
rs121907981 Chr.15: 72345550-72345550 on Build GRCh38
rs111827252 Chr.15: 72345209-72345209 on Build GRCh38
rs4777502 Chr.15: 72345454-72345454 on Build GRCh38
rs944022741 Chr.15: 72345125-72345125 on Build GRCh38
rs957797385 Chr.15: 72345273-72345273 on Build GRCh38
rs1022234819 Chr.15: 72345323-72345323 on Build GRCh38
rs376929315 Chr.15: 72345533-72345533 on Build GRCh38
rs149948017 Chr.15: 72345382-72345382 on Build GRCh38
rs1057519467 Chr.15: 72345540-72345540 on Build GRCh38
rs545914869 Chr.15: 72345273-72345273 on Build GRCh38
rs376303856 Chr.15: 72345416-72345416 on Build GRCh38
rs121907966 Chr.15: 72345477-72345477 on Build GRCh38
rs1016244855 Chr.15: 72345238-72345238 on Build GRCh38
rs766483588 Chr.15: 72345524-72345524 on Build GRCh38
rs145012038 Chr.15: 72345537-72345537 on Build GRCh38
rs1019828065 Chr.15: 72345343-72345343 on Build GRCh38
rs147502219 Chr.15: 72345482-72345482 on Build GRCh38
rs1038206117 Chr.15: 72345342-72345342 on Build GRCh38
rs886509268 Chr.15: 72345334-72345334 on Build GRCh38
rs1030815417 Chr.15: 72345391-72345391 on Build GRCh38
rs200180873 Chr.15: 72345417-72345417 on Build GRCh38
rs370638338 Chr.15: 72345534-72345534 on Build GRCh38
rs370449921 Chr.15: 72345434-72345434 on Build GRCh38
rs376373924 Chr.15: 72345424-72345424 on Build GRCh38
rs900521854 Chr.15: 72345255-72345255 on Build GRCh38
rs999341128 Chr.15: 72345389-72345389 on Build GRCh38
rs376576075 Chr.15: 72345316-72345316 on Build GRCh38
rs537902980 Chr.15: 72345127-72345127 on Build GRCh38
rs923723129 Chr.15: 72345058-72345058 on Build GRCh38
rs121907968 Chr.15: 72345519-72345519 on Build GRCh38
rs774313739 Chr.15: 72345495-72345495 on Build GRCh38
rs370091400 Chr.15: 72345377-72345377 on Build GRCh38
rs977951628 Chr.15: 72345272-72345273 on Build GRCh38
rs2912218 Chr.15: 72345367-72345367 on Build GRCh38
rs989247865 Chr.15: 72345294-72345294 on Build GRCh38
rs887606719 Chr.15: 72345210-72345210 on Build GRCh38
rs1044298723 Chr.15: 72345241-72345241 on Build GRCh38
rs912571796 Chr.15: 72345089-72345089 on Build GRCh38
rs757609740 Chr.15: 72345423-72345423 on Build GRCh38
rs891448877 Chr.15: 72345486-72345486 on Build GRCh38
rs140091006 Chr.15: 72345529-72345529 on Build GRCh38
rs528390276 Chr.15: 72345118-72345119 on Build GRCh38
rs121907952 Chr.15: 72345528-72345528 on Build GRCh38

View Full Product Details