The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003019.4 | NP_003010.4 | 3 - 4 | 354 | 109 |
XM_011540087.1 | XP_011538389.1 | 3 - 4 | 333 | 109 | |
GenBank mRNA | BC022318.1 | - | 3 - 4 | 355 | 109 |
BI820265.1 | - | 2 - 3 | 326 | 109 | |
CR541948.1 | - | 2 - 3 | 311 | 109 | |
X65018.1 | - | 3 - 4 | 482 | 109 |