The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014227.2 | NP_055042.1 | 1 - 2 | 139 | 62 |
GenBank mRNA | AJ133127.1 | - | 1 - 2 | 153 | 62 |