The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080.3 | NP_001071.1 | 2 - 3 | 470 | 67 |
NM_170740.1 | NP_733936.1 | 2 - 3 | 470 | 67 | |
GenBank mRNA | AJ427353.1 | - | 2 - 3 | 238 | 67 |
AJ427354.2 | - | 2 - 3 | 238 | 67 | |
AJ427355.2 | - | 2 - 3 | 442 | 67 | |
AK290800.1 | - | 2 - 3 | 588 | 67 | |
AK294699.1 | - | 3 - 4 | 247 | 67 | |
AK314357.1 | - | 2 - 3 | 470 | 67 | |
AK315380.1 | - | 2 - 3 | 565 | 67 | |
BC034321.1 | - | 2 - 3 | 470 | 67 | |
KJ534767.1 | - | 2 - 3 | 442 | 67 | |
KJ534768.1 | - | 2 - 3 | 442 | 67 | |
KJ534769.1 | - | 2 - 3 | 442 | 67 | |
Y11192.1 | - | 2 - 3 | 442 | 67 |