The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173598.4 | NP_775869.3 | 6 - 7 | 1206 | 75 |
XM_011538224.2 | XP_011536526.1 | 6 - 7 | 1541 | 75 | |
XM_011538225.2 | XP_011536527.1 | 6 - 7 | 2000 | 75 | |
XM_011538226.2 | XP_011536528.1 | 6 - 7 | 1541 | 75 | |
XM_011538228.2 | XP_011536530.1 | 4 - 5 | 365 | 75 | |
XM_011538229.2 | XP_011536531.1 | 6 - 7 | 1541 | 75 | |
XM_017019208.1 | XP_016874697.1 | 6 - 7 | 1541 | 75 | |
XM_017019209.1 | XP_016874698.1 | 6 - 7 | 1541 | 75 | |
GenBank mRNA | AK098831.1 | - | 4 - 5 | 384 | 75 |
AY345972.1 | - | 4 - 5 | 875 | 75 | |
BC107106.1 | - | 4 - 5 | 870 | 75 | |
BC107107.1 | - | 4 - 5 | 870 | 75 | |
BC127603.1 | - | 4 - 5 | 875 | 75 |