The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002754.4 | NP_002745.1 | 1 - 2 | 217 | 88 |
NR_072996.1 | 1 - 2 | 217 | 88 | ||
GenBank mRNA | AB451221.1 | - | 1 - 3 | 88 | |
AB451344.1 | - | 1 - 3 | 88 | ||
AF015256.1 | - | 1 - 2 | 178 | 88 | |
AF092535.1 | - | 1 - 2 | 139 | 88 | |
AF100546.1 | - | 1 - 2 | 120 | 88 | |
AK222922.1 | - | 1 - 2 | 109 | 88 | |
AK308018.1 | - | 1 - 2 | 198 | 88 | |
BC000433.2 | - | 1 - 2 | 159 | 88 | |
BC001641.2 | - | 1 - 2 | 159 | 88 | |
BC004428.1 | - | 1 - 2 | 159 | 88 | |
BC085196.1 | - | 1 - 2 | 187 | 88 | |
BM921729.1 | - | 1 - 2 | 216 | 88 | |
BT007221.1 | - | 1 - 2 | 118 | 88 | |
CR536490.1 | - | 1 - 2 | 118 | 88 | |
DB473379.1 | - | 1 - 2 | 181 | 88 | |
DR006449.1 | - | 1 - 2 | 221 | 88 | |
KU178281.1 | - | 1 - 2 | 118 | 88 | |
KU178282.1 | - | 1 - 2 | 118 | 88 | |
U93232.1 | - | 1 - 2 | 125 | 88 | |
Y10488.1 | - | 1 - 2 | 140 | 88 |