Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLR8
Entrez Gene ID
51311
Gene Name
toll like receptor 8
Gene Aliases
CD288
Chromosome Location
Chr.X: 12906620 - 12923169 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138636.5 NP_619542.1 1 - 2 88 106
GenBank mRNA AF245703.1 - 1 - 2 49 106
AK314240.1 - 1 - 2 88 106
AY358296.1 - 1 - 2 57 106
BC101074.1 - 1 - 2 55 106
BC101075.2 - 1 - 2 55 106
BC101076.2 - 1 - 2 55 106
BC101077.1 - 1 - 2 55 106

View Full Product Details