The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000673.4 | NP_000664.2 | 3 - 4 | 540 | 101 |
NM_001166504.1 | NP_001159976.1 | 3 - 4 | 353 | 101 | |
GenBank mRNA | AK223472.1 | - | 3 - 4 | 334 | 101 |
AK301696.1 | - | 3 - 4 | 353 | 101 | |
AK312854.1 | - | 3 - 4 | 463 | 101 | |
BC131512.1 | - | 3 - 4 | 335 | 101 | |
BX476080.1 | - | 3 - 4 | 397 | 101 | |
CD014081.1 | - | 4 - 5 | 416 | 101 | |
L33179.1 | - | 3 - 4 | 263 | 101 | |
U07821.1 | - | 3 - 4 | 347 | 101 | |
U09623.1 | - | 3 - 4 | 281 | 101 | |
X76342.1 | - | 3 - 4 | 323 | 101 |