The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004747.3 | NP_004738.3 | 18 - 19 | 3855 | 104 |
XM_005270276.4 | XP_005270333.1 | 18 - 19 | 3809 | 104 | |
XM_006718056.3 | XP_006718119.1 | 17 - 18 | 2798 | 104 | |
XM_006718057.3 | XP_006718120.1 | 18 - 19 | 3825 | 104 | |
XM_011540341.2 | XP_011538643.1 | 18 - 19 | 3979 | 104 | |
XM_011540342.1 | XP_011538644.1 | 18 - 19 | 3557 | 104 | |
XM_011540344.2 | XP_011538646.1 | 17 - 18 | 3464 | 104 | |
XM_011540345.1 | XP_011538647.1 | 17 - 18 | 3589 | 104 | |
XM_011540346.2 | XP_011538648.1 | 18 - 19 | 3824 | 104 | |
XM_011540347.2 | XP_011538649.1 | 14 - 15 | 3035 | 104 | |
XM_017016913.1 | XP_016872402.1 | 18 - 19 | 3518 | 104 | |
XM_017016914.1 | XP_016872403.1 | 18 - 19 | 3848 | 104 | |
XM_017016915.1 | XP_016872404.1 | 14 - 15 | 3719 | 104 | |
GenBank mRNA | AB011155.2 | - | 18 - 19 | 3885 | 104 |
AB091676.1 | - | 16 - 17 | 3320 | 104 | |
AK055172.1 | - | 14 - 15 | 2773 | 104 | |
AK122931.1 | - | 19 - 20 | 3787 | 104 | |
AK314771.1 | - | 2 - 3 | 164 | 104 | |
BC073996.1 | - | 4 - 5 | 565 | 104 | |
BC117695.2 | - | 8 - 9 | 3613 | 104 | |
BC146794.1 | - | 18 - 19 | 3885 | 104 | |
BX649131.1 | - | 13 - 14 | 1962 | 104 | |
U61843.1 | - | 3 - 4 | 413 | 104 |