Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72349100-72349331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
UniGene
Hs.604479
Gene Chromosome Location
Chr.15: 72343435-72376473 on Build GRCh38
Transcripts
NM_000520.5 NM_001318825.1 NR_134869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6219028 c.947A>C p.Y316S Chr.15: 72349118-72349118 on Build GRCh38
98316 c.856A>G p.T286A Chr.15: 72349209-72349209 on Build GRCh38
1254146 c.816A>T p.G272G Chr.15: 72349249-72349249 on Build GRCh38
4509569 c.810C>T p.I270I Chr.15: 72349255-72349255 on Build GRCh38
701309 c.912C>G p.F304L Chr.15: 72349153-72349153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765483440 Chr.15: 72349292-72349292 on Build GRCh38
rs199578185 Chr.15: 72349181-72349181 on Build GRCh38
rs1057519463 Chr.15: 72349160-72349167 on Build GRCh38
rs121907977 Chr.15: 72349163-72349163 on Build GRCh38
rs768518776 Chr.15: 72349210-72349210 on Build GRCh38
rs778191188 Chr.15: 72349169-72349169 on Build GRCh38
rs780105682 Chr.15: 72349158-72349168 on Build GRCh38
rs752260651 Chr.15: 72349177-72349177 on Build GRCh38
rs773093433 Chr.15: 72349239-72349239 on Build GRCh38
rs776785471 Chr.15: 72349282-72349282 on Build GRCh38
rs772036483 Chr.15: 72349232-72349232 on Build GRCh38
rs758871181 Chr.15: 72349164-72349164 on Build GRCh38
rs779688204 Chr.15: 72349194-72349194 on Build GRCh38
rs911351010 Chr.15: 72349250-72349250 on Build GRCh38
rs751248523 Chr.15: 72349135-72349136 on Build GRCh38
rs763364823 Chr.15: 72349305-72349305 on Build GRCh38
rs775809111 Chr.15: 72349303-72349303 on Build GRCh38
rs759837903 Chr.15: 72349158-72349158 on Build GRCh38
rs747046058 Chr.15: 72349262-72349262 on Build GRCh38
rs190019714 Chr.15: 72349287-72349287 on Build GRCh38
rs121907960 Chr.15: 72349153-72349155 on Build GRCh38
rs1054374 Chr.15: 72349187-72349187 on Build GRCh38
rs143009489 Chr.15: 72349189-72349189 on Build GRCh38
rs758406312 Chr.15: 72349156-72349156 on Build GRCh38
rs748958444 Chr.15: 72349207-72349207 on Build GRCh38
rs73440586 Chr.15: 72349307-72349307 on Build GRCh38
rs770932296 Chr.15: 72349266-72349266 on Build GRCh38
rs763685631 Chr.15: 72349311-72349311 on Build GRCh38
rs778823870 Chr.15: 72349225-72349225 on Build GRCh38
rs1051875886 Chr.15: 72349310-72349310 on Build GRCh38
rs148199798 Chr.15: 72349229-72349229 on Build GRCh38
rs778155650 Chr.15: 72349148-72349150 on Build GRCh38
rs765612435 Chr.15: 72349159-72349159 on Build GRCh38
rs1057516908 Chr.15: 72349260-72349260 on Build GRCh38

View Full Product Details