Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1349775-1350020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UVSSA
Entrez Gene ID
57654
Gene Name
UV stimulated scaffold protein A
Gene Aliases
KIAA1530, UVSS3
UniGene
Hs.380475
Gene Chromosome Location
Chr.4: 1345662-1388049 on Build GRCh38
Transcripts
NM_001317934.1 NM_001317935.1 NM_020894.3 XM_017008490.1 XM_017008491.1 XM_017008492.1 XM_017008493.1 XM_017008494.1 XM_017008496.1 XM_017008497.1 XM_017008498.1 XM_017008499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
293548 c.372_373insG p.E126fs*44 Chr.4: 1349797-1349798 on Build GRCh38
3775558 c.355G>A p.G119R Chr.4: 1349780-1349780 on Build GRCh38
1051246 c.425A>C p.K142T Chr.4: 1349850-1349850 on Build GRCh38
5666664 c.356G>T p.G119V Chr.4: 1349781-1349781 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377071239 Chr.4: 1349821-1349821 on Build GRCh38
rs775003416 Chr.4: 1349839-1349839 on Build GRCh38
rs935790720 Chr.4: 1349880-1349880 on Build GRCh38
rs760103572 Chr.4: 1349849-1349849 on Build GRCh38
rs940029830 Chr.4: 1349936-1349936 on Build GRCh38
rs771676698 Chr.4: 1349837-1349837 on Build GRCh38
rs748234387 Chr.4: 1349799-1349799 on Build GRCh38
rs199674866 Chr.4: 1349866-1349866 on Build GRCh38
rs898555865 Chr.4: 1349947-1349947 on Build GRCh38
rs549702058 Chr.4: 1349960-1349960 on Build GRCh38
rs751989306 Chr.4: 1349863-1349863 on Build GRCh38
rs887083163 Chr.4: 1349934-1349934 on Build GRCh38
rs768698987 Chr.4: 1349880-1349881 on Build GRCh38
rs369196034 Chr.4: 1349809-1349809 on Build GRCh38
rs749042254 Chr.4: 1349812-1349812 on Build GRCh38
rs541124924 Chr.4: 1349946-1349946 on Build GRCh38
rs377140234 Chr.4: 1349836-1349836 on Build GRCh38
rs146386291 Chr.4: 1349931-1349931 on Build GRCh38
rs760108046 Chr.4: 1349916-1349916 on Build GRCh38
rs888954591 Chr.4: 1349989-1349989 on Build GRCh38
rs561542353 Chr.4: 1350000-1350000 on Build GRCh38
rs765738301 Chr.4: 1349852-1349852 on Build GRCh38
rs1028774938 Chr.4: 1349985-1349985 on Build GRCh38
rs372688958 Chr.4: 1349833-1349833 on Build GRCh38
rs185686321 Chr.4: 1349893-1349893 on Build GRCh38
rs1008726644 Chr.4: 1349998-1349998 on Build GRCh38
rs763503368 Chr.4: 1349858-1349858 on Build GRCh38
rs766699529 Chr.4: 1349859-1349859 on Build GRCh38
rs778046949 Chr.4: 1349898-1349898 on Build GRCh38
rs572514109 Chr.4: 1349899-1349899 on Build GRCh38
rs367917115 Chr.4: 1349886-1349886 on Build GRCh38
rs750755892 Chr.4: 1349953-1349953 on Build GRCh38
rs770825185 Chr.4: 1349830-1349830 on Build GRCh38
rs937712661 Chr.4: 1349912-1349912 on Build GRCh38
rs755215063 Chr.4: 1349885-1349885 on Build GRCh38
rs372165812 Chr.4: 1349897-1349897 on Build GRCh38
rs769715188 Chr.4: 1349806-1349806 on Build GRCh38
rs112417076 Chr.4: 1349814-1349814 on Build GRCh38
rs750954143 Chr.4: 1349857-1349857 on Build GRCh38
rs1000694185 Chr.4: 1349983-1349983 on Build GRCh38
rs11247994 Chr.4: 1349932-1349932 on Build GRCh38
rs1020677822 Chr.4: 1349952-1349952 on Build GRCh38

View Full Product Details