Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88396057-88396316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA7
Entrez Gene ID
55812
Gene Name
spermatogenesis associated 7
Gene Aliases
HEL-S-296, HSD-3.1, HSD3, LCA3
UniGene
Hs.525518
Gene Chromosome Location
Chr.14: 88385638-88470350 on Build GRCh38
Transcripts
NM_001040428.3 NM_018418.4 XM_005267851.1 XM_005267852.1 XM_005267854.1 XM_005267855.1 XM_006720204.1 XM_006720205.1 XM_011536951.1 XM_011536952.1 XM_011536953.1 XM_017021452.1 XM_017021453.1 XM_017021454.1 XM_017021455.1 XM_017021456.1 XM_017021457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3936646 c.223A>T p.S75C Chr.14: 88396188-88396188 on Build GRCh38
1227284 c.226A>G p.T76A Chr.14: 88396191-88396191 on Build GRCh38
7223236 c.206C>T p.S69L Chr.14: 88396171-88396171 on Build GRCh38
958433 c.204C>T p.C68C Chr.14: 88396169-88396169 on Build GRCh38
7234862 c.220G>A p.V74M Chr.14: 88396185-88396185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774821432 Chr.14: 88396159-88396159 on Build GRCh38
rs764345380 Chr.14: 88396150-88396150 on Build GRCh38
rs951511456 Chr.14: 88396098-88396098 on Build GRCh38
rs781049303 Chr.14: 88396219-88396219 on Build GRCh38
rs3179969 Chr.14: 88396185-88396185 on Build GRCh38
rs751477368 Chr.14: 88396221-88396223 on Build GRCh38
rs773899103 Chr.14: 88396240-88396240 on Build GRCh38
rs762300671 Chr.14: 88396165-88396165 on Build GRCh38
rs149093540 Chr.14: 88396235-88396235 on Build GRCh38
rs142913613 Chr.14: 88396172-88396172 on Build GRCh38
rs772573394 Chr.14: 88396250-88396250 on Build GRCh38
rs930835760 Chr.14: 88396104-88396104 on Build GRCh38
rs1049238392 Chr.14: 88396108-88396108 on Build GRCh38
rs768088420 Chr.14: 88396167-88396167 on Build GRCh38
rs745808329 Chr.14: 88396221-88396221 on Build GRCh38
rs745332612 Chr.14: 88396126-88396126 on Build GRCh38
rs775122024 Chr.14: 88396227-88396227 on Build GRCh38
rs748863676 Chr.14: 88396234-88396234 on Build GRCh38
rs563947621 Chr.14: 88396119-88396119 on Build GRCh38
rs371770633 Chr.14: 88396152-88396152 on Build GRCh38
rs754983533 Chr.14: 88396235-88396235 on Build GRCh38
rs757341191 Chr.14: 88396118-88396121 on Build GRCh38
rs753995098 Chr.14: 88396192-88396192 on Build GRCh38
rs777396619 Chr.14: 88396209-88396209 on Build GRCh38
rs757062701 Chr.14: 88396212-88396212 on Build GRCh38
rs531843956 Chr.14: 88396279-88396279 on Build GRCh38
rs762732339 Chr.14: 88396147-88396147 on Build GRCh38
rs373094088 Chr.14: 88396177-88396177 on Build GRCh38
rs927558911 Chr.14: 88396096-88396096 on Build GRCh38
rs371901116 Chr.14: 88396138-88396138 on Build GRCh38
rs769306535 Chr.14: 88396134-88396134 on Build GRCh38
rs1041333614 Chr.14: 88396197-88396197 on Build GRCh38
rs60770744 Chr.14: 88396142-88396142 on Build GRCh38
rs996186010 Chr.14: 88396178-88396178 on Build GRCh38
rs1050146507 Chr.14: 88396181-88396181 on Build GRCh38
rs756199454 Chr.14: 88396184-88396184 on Build GRCh38
rs766336763 Chr.14: 88396217-88396217 on Build GRCh38
rs958554500 Chr.14: 88396161-88396161 on Build GRCh38
rs755274487 Chr.14: 88396204-88396204 on Build GRCh38
rs563469560 Chr.14: 88396090-88396090 on Build GRCh38
rs143184396 Chr.14: 88396242-88396242 on Build GRCh38
rs769180883 Chr.14: 88396222-88396222 on Build GRCh38
rs569231437 Chr.14: 88396077-88396077 on Build GRCh38
rs148283835 Chr.14: 88396271-88396271 on Build GRCh38
rs781090403 Chr.14: 88396234-88396235 on Build GRCh38

View Full Product Details