Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CATIP
Entrez Gene ID
375307
Gene Name
ciliogenesis associated TTC17 interacting protein
Gene Aliases
C2orf62
Chromosome Location
Chr.2: 218356757 - 218368393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320865.1 NP_001307794.1 4 - 5 439 63
NM_198559.1 NP_940961.1 4 - 5 406 63
XM_005246539.4 XP_005246596.1 3 - 4 403 63
XM_005246541.4 XP_005246598.1 2 - 3 202 63
XM_011511148.2 XP_011509450.1 4 - 5 538 63
XM_011511149.2 XP_011509451.1 2 - 3 201 63
XM_011511150.2 XP_011509452.1 2 - 3 167 63
XM_017004053.1 XP_016859542.1 2 - 3 574 63
GenBank mRNA BC052750.1 - 4 - 5 406 63

View Full Product Details