The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006401.2 | NP_006392.1 | 7 - 7 | 1214 | 158 |
GenBank mRNA | BC019658.1 | - | 7 - 7 | 1053 | 158 |
U70439.1 | - | 7 - 7 | 1075 | 158 | |
Y07570.1 | - | 6 - 6 | 881 | 158 | |
Y07969.1 | - | 7 - 7 | 1087 | 156 |