The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142929.1 | NP_001136401.1 | 12 - 13 | 1932 | 64 |
NM_001142933.1 | NP_001136405.1 | 13 - 14 | 2002 | 64 | |
NM_001142934.1 | NP_001136406.1 | 13 - 14 | 2002 | 64 | |
NM_020549.4 | NP_065574.3 | 12 - 13 | 1932 | 64 | |
NM_020984.3 | NP_066264.3 | 12 - 13 | 1598 | 64 | |
NM_020985.3 | NP_066265.3 | 13 - 14 | 1760 | 64 | |
NM_020986.3 | NP_066266.3 | 12 - 13 | 1588 | 64 | |
GenBank mRNA | AF305907.1 | - | 12 - 13 | 1932 | 64 |
AF305908.2 | - | 13 - 14 | 2002 | 64 | |
AF305909.1 | - | 12 - 13 | 1598 | 64 | |
BC130615.1 | - | 11 - 12 | 1458 | 64 | |
BC130617.1 | - | 11 - 12 | 1458 | 64 | |
S45018.1 | - | 11 - 12 | 1449 | 64 | |
S56138.1 | - | 12 - 13 | 1932 | 64 |